Assay Details
| Gene Symbol: SYNRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005256980.4 | XP_005257037.1 | 7 - 8 | 1034 | 92 | |
| XM_017024076.1 | XP_016879565.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024077.1 | XP_016879566.1 | 7 - 8 | 953 | 92 | ||
| XM_017024078.1 | XP_016879567.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024079.1 | XP_016879568.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024080.1 | XP_016879569.1 | 7 - 8 | 5086 | 92 | ||
| XM_017024081.1 | XP_016879570.1 | 7 - 8 | 5083 | 92 | ||
| XM_017024082.1 | XP_016879571.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024083.1 | XP_016879572.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024084.1 | XP_016879573.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024085.1 | XP_016879574.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024086.1 | XP_016879575.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024087.1 | XP_016879576.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024089.1 | XP_016879578.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024092.1 | XP_016879581.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024096.1 | XP_016879585.1 | 7 - 8 | 1034 | 92 | ||
| XM_017024105.1 | XP_016879594.1 | 7 - 8 | 1034 | 92 | ||
| GenBank mRNA | AK302821.1 | - | 7 - 8 | 978 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SYNRG
Genomic Map