| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030227.hg.1 |
ENST00000366578.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030227.hg.1 |
ENST00000492634.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030227.hg.1 |
ENST00000542672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030227.hg.1 |
ENST00000545611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030227.hg.1 |
NM_001103.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030227.hg.1 |
TC01001955.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
ENST00000366578.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
ENST00000492634.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
ENST00000542672.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
ENST00000545611.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
ENST00000546208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
NM_001103.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030226.hg.1 |
TC01001955.hg.1 |
HTA TC |