Assay Details
| Gene Symbol: TRPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251845.1 | NP_001238774.1 | 3 - 4 | 557 | 134 | |
| XM_005247738.3 | XP_005247795.1 | 2 - 3 | 232 | 134 | ||
| XM_017007121.1 | XP_016862610.1 | 3 - 4 | 1165 | 134 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
manganese ion transport
melanin biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
regulation of cytosolic calcium ion concentration
response to calcium ion
calcium ion transmembrane transport
cellular response to hypoxia
regulation of cardiac conduction
cation channel activity
calcium channel activity
protein binding
store-operated calcium channel activity
inositol 1,4,5 trisphosphate binding
calcium ion transport
manganese ion transport
melanin biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
regulation of cytosolic calcium ion concentration
response to calcium ion
calcium ion transmembrane transport
cellular response to hypoxia
regulation of cardiac conduction
cation channel activity
calcium channel activity
protein binding
store-operated calcium channel activity
inositol 1,4,5 trisphosphate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000460401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000460401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000460401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000480101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000480101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000480101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000273482.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000460401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000460401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000460401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000476941.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000480101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000480101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000480101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | ENST00000612385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_001251845.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009035.hg.1 | NM_003304.4 | Refseq |

TRPC1
Genomic Map