Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017008378.1 | XP_016863867.1 | 3 - 4 | 521 | 185 | |
| XM_017008379.1 | XP_016863868.1 | 3 - 4 | 516 | 185 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017008378.1, XM_017008379.1 transcripts.
Set Membership: |
|

AP1AR
Genomic Map