Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128426.2 | NP_001121898.1 | 5 - 6 | 637 | 99 | |
| XM_017008379.1 | XP_016863868.1 | 6 - 7 | 791 | 99 | ||
| GenBank mRNA | BC009485.2 | - | 5 - 6 | 543 | 3623656 | 99 |
More Information
Set Membership: |
|

AP1AR
Genomic Map