Assay Details
| Gene Symbol: LRRC4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080457.1 | NP_001073926.1 | 1 - 2 | 97 | 85 | |
| GenBank mRNA | BC131897.1 | - | 1 - 2 | 75 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001750.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC131897.1,NM_001080457.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19023814.hg.1 | ENST00000389201.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023814.hg.1 | ENST00000535879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023814.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023814.hg.1 | TC19001750.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023816.hg.1 | ENST00000389201.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023816.hg.1 | ENST00000535879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023816.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023816.hg.1 | TC19001750.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023813.hg.1 | ENST00000389201.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023813.hg.1 | ENST00000535879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023813.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023813.hg.1 | TC19001750.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023815.hg.1 | ENST00000389201.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023815.hg.1 | ENST00000535879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023815.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023815.hg.1 | TC19001750.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | TC1900011209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194145.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | TC1900011209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194146.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | TC1900011209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194147.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | TC1900011209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194149.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000389201.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000599957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | ENST00000600381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | NM_001080457.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011209.hg.1 | NM_001080457.1 | Refseq |

LRRC4B
Genomic Map