Assay Details
| Gene Symbol: KCNH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033272.3 | NP_150375.2 | 4 - 5 | 1123 | 98 | |
| XM_011512109.2 | XP_011510411.1 | 4 - 5 | 1013 | 122 | ||
| XM_017005218.1 | XP_016860707.1 | 4 - 5 | 1013 | 122 | ||
| XM_017005219.1 | XP_016860708.1 | 4 - 5 | 1013 | 98 | ||
| XM_017005221.1 | XP_016860710.1 | 4 - 5 | 1013 | 122 | ||
| GenBank mRNA | AF032897.1 | - | 4 - 5 | 994 | 98 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |

KCNH7
Genomic Map