| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008591.hg.1 |
ENST00000355540.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008591.hg.1 |
ENST00000359863.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008591.hg.1 |
ENST00000446306.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008591.hg.1 |
NM_001244580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008591.hg.1 |
NM_003496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07008591.hg.1 |
TC07003396.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000355540.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000359863.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000446306.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000456197.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000628380.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
NM_001244580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
NM_003496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000355540.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000359863.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000446306.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000456197.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
ENST00000628380.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
NM_001244580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
NM_003496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700197861.hg.1 |
TC0700013413.hg.1 |
Clariom D TC |