| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000443095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000443095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
ENST00000443095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013139.hg.1 |
ENST00000344924.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013139.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013139.hg.1 |
ENST00000418040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013139.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013139.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013139.hg.1 |
TC07000836.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013136.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013136.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07013136.hg.1 |
TC07000836.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165621.hg.1 |
TC0700009232.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700165618.hg.1 |
TC0700009232.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000344924.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000393132.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000418040.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000443095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000443095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
ENST00000443095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001293085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_001724.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700009232.hg.1 |
NM_199186.2 |
Refseq |