Assay Details
| Gene Symbol: ARNTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030273.2 | NP_001025444.1 | 5 - 6 | 617 | 122 | |
| NM_001297724.1 | NP_001284653.1 | 5 - 6 | 610 | 122 | ||
| XM_006718234.1 | XP_006718297.1 | 5 - 6 | 554 | 122 | ||
| XM_011520110.1 | XP_011518412.1 | 6 - 7 | 609 | 122 | ||
| XM_011520111.1 | XP_011518413.1 | 4 - 5 | 424 | 122 | ||
| XM_011520112.2 | XP_011518414.1 | 6 - 7 | 609 | 122 | ||
| XM_011520113.1 | XP_011518415.1 | 6 - 7 | 609 | 122 | ||
| XM_017017744.1 | XP_016873233.1 | 6 - 7 | 609 | 122 | ||
| XM_017017745.1 | XP_016873234.1 | 5 - 6 | 554 | 122 | ||
| XM_017017746.1 | XP_016873235.1 | 5 - 6 | 564 | 122 | ||
| XM_017017747.1 | XP_016873236.1 | 4 - 5 | 424 | 122 | ||
| XM_017017748.1 | XP_016873237.1 | 4 - 5 | 481 | 122 | ||
| GenBank mRNA | AB000815.1 | - | 3 - 4 | 253 | 122 | |
| AB000816.1 | - | 4 - 5 | 593 | 122 | ||
| AK095749.1 | - | 5 - 6 | 502 | 122 | ||
| BC016674.1 | - | 2 - 3 | 145 | 3921870 | 122 | |
| BC031214.1 | - | 5 - 6 | 442 | 4793007 | 122 | |
| D89722.1 | - | 2 - 3 | 158 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB000815.1, AB000816.1, AK095749.1, BC016674.1, BC031214.1, D89722.1, NM_001030273.2, NM_001297724.1, XM_006718234.1, XM_011520110.1, XM_011520111.1, XM_011520112.2, XM_011520113.1, XM_017017744.1, XM_017017745.1, XM_017017746.1, XM_017017747.1, XM_017017748.1 transcripts.
Set Membership: |
|

ARNTL
Genomic Map