Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135579.1 | 7 - 8 | 2179 | 121 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000362063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000430262.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000508237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | ENST00000513488.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_001145365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011038.hg.1 | NM_014897.2 | Refseq |

ZNF652
Genomic Map