Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018181.5 | NP_060651.2 | 2 - 3 | 651 | 136 | |
| XM_017025810.1 | XP_016881299.1 | 2 - 3 | 651 | 136 | ||
| GenBank mRNA | AF295077.1 | - | 2 - 3 | 436 | 136 | |
| AK128058.1 | - | 2 - 3 | 653 | 136 | ||
| BC130620.1 | - | 2 - 3 | 303 | 40146651 | 136 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK128058.1, BC130620.1, NM_018181.5, XM_017025810.1 transcripts.
Set Membership: |
|

ZNF532
Genomic Map