Assay Details
| Gene Symbol: NOL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317724.1 | NP_001304653.1 | 8 - 9 | 841 | 103 | |
| GenBank mRNA | BU071935.1 | - | 8 - 9 | 594 | 103 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000420088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000451315.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | ENST00000474485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006993.hg.1 | NM_016167.4 | Refseq |

NOL7
Genomic Map