| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024567.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024567.hg.1 |
ENST00000512158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024567.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024567.hg.1 |
TC05001804.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024569.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024569.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024569.hg.1 |
TC05001804.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024565.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024565.hg.1 |
ENST00000512158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024565.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024565.hg.1 |
TC05001804.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024568.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024568.hg.1 |
ENST00000512158.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024568.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024568.hg.1 |
TC05001804.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012107.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183800.hg.1 |
TC0500012107.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500183802.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183802.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183802.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183802.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183802.hg.1 |
TC0500012107.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183799.hg.1 |
TC0500012107.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183798.hg.1 |
TC0500012107.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
ENST00000337225.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
ENST00000512158.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
NM_004887.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183801.hg.1 |
TC0500012107.hg.1 |
Clariom D TC |