| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013739.hg.1 |
ENST00000310917.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013739.hg.1 |
ENST00000344408.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013739.hg.1 |
ENST00000344938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013739.hg.1 |
NM_001166136.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013739.hg.1 |
NM_147127.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013739.hg.1 |
TC04000996.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000310917.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000344408.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000475313.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000509670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
NM_001166136.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
NM_147127.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000310917.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000344408.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000475313.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
ENST00000509670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
NM_001166136.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
NM_147127.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167997.hg.1 |
TC0400009891.hg.1 |
Clariom D TC |