Assay Details
| Gene Symbol: GLP2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC019818.1 | - | 6 - 7 | 614 | 64 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000262441.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000304773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000458005.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | ENST00000574745.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006860.hg.1 | NM_004246.2 | Refseq |

GLP2R
Genomic Map