| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
ENST00000231526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
ENST00000399438.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
ENST00000438419.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
ENST00000505553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
ENST00000545191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
NM_001093755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
NM_001093756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
NM_001243737.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
NM_024941.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05003489.hg.1 |
TC05000278.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000231526.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000399438.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000415825.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000438419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000505108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000505553.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000509234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000512009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_001093755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_001093756.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_001243737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_024941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000231526.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000399438.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000415825.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000438419.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000505108.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000505553.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000509234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
ENST00000512009.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_001093755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_001093756.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_001243737.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
NM_024941.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500151338.hg.1 |
TC0500007605.hg.1 |
Clariom D TC |