| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050611.hg.1 |
ENST00000271854.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050611.hg.1 |
ENST00000368553.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050611.hg.1 |
ENST00000368559.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050611.hg.1 |
NM_001159484.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050611.hg.1 |
NM_207308.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050611.hg.1 |
TC01003284.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
ENST00000271854.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
ENST00000368553.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
ENST00000368559.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
NM_001159484.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
NM_207308.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
ENST00000271854.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
ENST00000368553.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
ENST00000368559.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
NM_001159484.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
NM_207308.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219592.hg.1 |
TC0100015896.hg.1 |
Clariom D TC |