Assay Details
| Gene Symbol: COMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000095.2 | NP_000086.2 | 6 - 7 | 638 | 77 | |
| GenBank mRNA | AB086984.1 | - | 6 - 7 | 602 | 77 | |
| AK074508.1 | - | 6 - 7 | 477 | 77 | ||
| AK290595.1 | - | 6 - 7 | 638 | 77 | ||
| AK296586.1 | - | 5 - 6 | 479 | 77 | ||
| AK297798.1 | - | 5 - 6 | 540 | 77 | ||
| AK297814.1 | - | 5 - 6 | 599 | 77 | ||
| BC033676.1 | - | 5 - 6 | 862 | 77 | ||
| BC110847.1 | - | 6 - 7 | 624 | 5591535 | 77 | |
| BC125092.1 | - | 6 - 7 | 631 | 40118200 | 77 | |
| L32137.1 | - | 6 - 7 | 627 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001293.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
growth plate cartilage development
apoptotic process
cell adhesion
organ morphogenesis
extracellular matrix organization
negative regulation of apoptotic process
limb development
protease binding
extracellular matrix structural constituent
calcium ion binding
protein binding
collagen binding
heparin binding
heparan sulfate proteoglycan binding
skeletal system development
growth plate cartilage development
apoptotic process
cell adhesion
organ morphogenesis
extracellular matrix organization
negative regulation of apoptotic process
limb development
protease binding
extracellular matrix structural constituent
calcium ion binding
protein binding
collagen binding
heparin binding
heparan sulfate proteoglycan binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19018392.hg.1 | ENST00000222271.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19018392.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19018392.hg.1 | ENST00000454701.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19018392.hg.1 | ENST00000542601.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19018392.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19018392.hg.1 | TC19001293.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | TC1900010031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900182447.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000222271.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000425807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000542601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | ENST00000612179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010031.hg.1 | NM_000095.2 | Refseq |

COMP
Genomic Map