Assay Details
| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509078.2 | XP_011507380.1 | 8 - 9 | 1034 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR3C| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509078.2 | XP_011507380.1 | 8 - 9 | 1034 | 118 | |
Set Membership: |
|