Assay Details
| Gene Symbol: NTNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113226.2 | NP_001106697.1 | 3 - 4 | 1585 | 79 | |
| XM_011541021.1 | XP_011539323.1 | 4 - 5 | 2501 | 79 | ||
| XM_017000680.1 | XP_016856169.1 | 4 - 5 | 1867 | 79 | ||
| XM_017000681.1 | XP_016856170.1 | 4 - 5 | 2820 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NTNG1
Genomic Map