Assay Details
| Gene Symbol: NCAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004540.3 | NP_004531.2 | 4 - 5 | 736 | 143 | |
| XM_011529575.2 | XP_011527877.1 | 5 - 6 | 1211 | 143 | ||
| XM_011529576.2 | XP_011527878.1 | 5 - 6 | 788 | 143 | ||
| XM_011529579.2 | XP_011527881.1 | 4 - 5 | 712 | 143 | ||
| XM_011529580.2 | XP_011527882.1 | 5 - 6 | 1211 | 143 | ||
| XM_011529581.2 | XP_011527883.1 | 5 - 6 | 1211 | 143 | ||
| XM_011529582.2 | XP_011527884.1 | 5 - 6 | 1210 | 143 | ||
| XM_017028353.1 | XP_016883842.1 | 5 - 6 | 876 | 143 | ||
| XM_017028354.1 | XP_016883843.1 | 4 - 5 | 712 | 143 | ||
| XM_017028355.1 | XP_016883844.1 | 4 - 5 | 712 | 143 | ||
| XM_017028358.1 | XP_016883847.1 | 5 - 6 | 1215 | 143 | ||
| GenBank mRNA | AK302870.1 | - | 5 - 6 | 715 | 143 | |
| BC052946.1 | - | 4 - 5 | 736 | 5315109 | 143 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000060.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21000290.hg.1 | ENST00000284894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000290.hg.1 | ENST00000400546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000290.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000290.hg.1 | ENST00000535285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000290.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000290.hg.1 | TC21000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | TC2100006714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145407.hg.1 | NM_004540.4 | Refseq |

NCAM2
Genomic Map