Assay Details
| Gene Symbol: IFI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002038.3 | NP_002029.3 | 1 - 2 | 84 | 95 | |
| NM_022872.2 | NP_075010.1 | 1 - 2 | 84 | 95 | ||
| NM_022873.2 | NP_075011.1 | 1 - 2 | 84 | 95 | ||
| GenBank mRNA | AK024814.1 | - | 1 - 2 | 64 | 95 | |
| AK314488.1 | - | 1 - 2 | 63 | 95 | ||
| BC011601.2 | - | 1 - 2 | 35 | 3140035 | 95 | |
| BC015603.2 | - | 1 - 2 | 37 | 4654295 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002412.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314488.1,BC011601.2,BC015603.2,NM_002038.3,NM_022872.2,NM_022873.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
release of cytochrome c from mitochondria
immune response
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of mitochondrial depolarization
type I interferon signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein binding
release of cytochrome c from mitochondria
immune response
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of mitochondrial depolarization
type I interferon signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | ENST00000339145.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | ENST00000361157.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037236.hg.1 | TC01002412.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | ENST00000339145.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | ENST00000361157.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037235.hg.1 | TC01002412.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199900.hg.1 | TC0100013445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199901.hg.1 | TC0100013445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013445.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | ENST00000339145.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | ENST00000361157.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | ENST00000362020.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | NM_002038.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | NM_022872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | NM_022873.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199902.hg.1 | TC0100013445.hg.1 | Clariom D TC |

IFI6
Genomic Map