| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
ENST00000339145.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
ENST00000361157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037231.hg.1 |
TC01002412.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
ENST00000339145.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
ENST00000361157.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037232.hg.1 |
TC01002412.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199897.hg.1 |
TC0100013445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199896.hg.1 |
TC0100013445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000339145.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000361157.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
ENST00000362020.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_002038.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022872.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013445.hg.1 |
NM_022873.2 |
Refseq |