Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152617.3 | NP_689830.2 | 5 - 6 | 1352 | 79 | |
| GenBank mRNA | AK054732.1 | - | 5 - 6 | 1352 | 79 | |
| AK093113.1 | - | 5 - 6 | 738 | 79 | ||
| BC017442.1 | - | 4 - 5 | 1184 | 79 | ||
| BC033791.1 | - | 5 - 6 | 1295 | 5163887 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002161.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
double-strand break repair via nonhomologous end joining
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
response to ionizing radiation
protein ubiquitination
protein sumoylation
negative regulation of transcription elongation from RNA polymerase II promoter
histone H2A monoubiquitination
interstrand cross-link repair
histone H2A-K13 ubiquitination
histone H2A-K15 ubiquitination
isotype switching
positive regulation of DNA repair
protein K63-linked ubiquitination
histone H2A K63-linked ubiquitination
chromatin binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
nucleosome binding
histone binding
ubiquitin binding
K63-linked polyubiquitin binding
double-strand break repair
double-strand break repair via nonhomologous end joining
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
response to ionizing radiation
protein ubiquitination
protein sumoylation
negative regulation of transcription elongation from RNA polymerase II promoter
histone H2A monoubiquitination
interstrand cross-link repair
histone H2A-K13 ubiquitination
histone H2A-K15 ubiquitination
isotype switching
positive regulation of DNA repair
protein K63-linked ubiquitination
histone H2A K63-linked ubiquitination
chromatin binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
nucleosome binding
histone binding
ubiquitin binding
K63-linked polyubiquitin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013702.hg.1 | ENST00000430043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038262.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038262.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038262.hg.1 | TC03002161.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038261.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038261.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038261.hg.1 | TC03002161.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202445.hg.1 | TC0300013703.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300202443.hg.1 | TC0300013703.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013702.hg.1 | ENST00000430043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000318037.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | ENST00000437070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013703.hg.1 | NM_152617.3 | Refseq |

RNF168
Genomic Map