| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
NM_002169.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
NM_002169.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009702.hg.1 |
NM_002169.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
ENST00000610521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
NM_002169.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
NM_002169.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009702.hg.1 |
NM_002169.2 |
Refseq |