| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01015851.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01015851.hg.1 |
ENST00000369630.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01015851.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01015851.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01015851.hg.1 |
TC01000993.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100168088.hg.1 |
TC0100009452.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000361886.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
ENST00000369630.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_001004440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009452.hg.1 |
NM_182759.2 |
Refseq |