Assay Details
| Gene Symbol: PRR14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040859.1 | - | 5 - 6 | 1085 | 5535580 | 113 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200161138.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200161138.hg.1 | TC2200008489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200161138.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000327423.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000330495.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000412743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000431684.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000432485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000461722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | ENST00000492705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008489.hg.1 | NM_173566.2 | Refseq |

PRR14L
Genomic Map