Assay Details
| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005199.4 | NP_005190.4 | 8 - 9 | 936 | 72 | |
| GenBank mRNA | AK125362.1 | - | 8 - 9 | 936 | 72 | |
| BC111802.1 | - | 7 - 8 | 759 | 40069605 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001403.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
muscle contraction
signal transduction
synaptic transmission, cholinergic
neuromuscular synaptic transmission
response to nicotine
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
protein binding
channel activity
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine binding
transport
muscle contraction
signal transduction
synaptic transmission, cholinergic
neuromuscular synaptic transmission
response to nicotine
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
protein binding
channel activity
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022160.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022160.hg.1 | ENST00000389494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022160.hg.1 | ENST00000541596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022160.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022160.hg.1 | TC02001403.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022159.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022159.hg.1 | ENST00000389494.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022159.hg.1 | ENST00000541596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022159.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022159.hg.1 | TC02001403.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179833.hg.1 | TC0200011105.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179834.hg.1 | TC0200011105.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389492.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000389494.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | ENST00000485094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011105.hg.1 | NM_005199.4 | Refseq |

CHRNG
Genomic Map