Assay Details
| Gene Symbol: ZNF335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022095.3 | NP_071378.1 | 11 - 12 | 1793 | 74 | |
| XM_005260504.4 | XP_005260561.1 | 11 - 12 | 2506 | 74 | ||
| XM_011528979.2 | XP_011527281.1 | 10 - 11 | 2161 | 74 | ||
| XM_017028012.1 | XP_016883501.1 | 10 - 11 | 1564 | 74 | ||
| GenBank mRNA | AF395833.1 | - | 11 - 12 | 1770 | 74 | |
| AK026157.1 | - | 3 - 4 | 308 | 74 | ||
| AK296989.1 | - | 8 - 9 | 1364 | 74 | ||
| BC041331.2 | - | 8 - 9 | 1008 | 74 | ||
| BC142981.1 | - | 8 - 9 | 1008 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000894.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
cerebral cortex neuron differentiation
regulation of gene expression, epigenetic
neuron projection morphogenesis
brain morphogenesis
positive regulation of lymphocyte proliferation
positive regulation of neurogenesis
regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
RNA polymerase II core promoter sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
in utero embryonic development
positive regulation of neuroblast proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
cerebral cortex neuron differentiation
regulation of gene expression, epigenetic
neuron projection morphogenesis
brain morphogenesis
positive regulation of lymphocyte proliferation
positive regulation of neurogenesis
regulation of histone H3-K4 methylation
histone H3-K4 trimethylation
RNA polymerase II core promoter sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20012214.hg.1 | ENST00000243961.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012214.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012214.hg.1 | ENST00000426788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012214.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012214.hg.1 | TC20000894.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000475002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000476822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | ENST00000494955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009253.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164788.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164788.hg.1 | NM_022095.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164788.hg.1 | TC2000009253.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164788.hg.1 | ENST00000322927.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164788.hg.1 | NM_022095.3 | Refseq |

ZNF335
Genomic Map