Assay Details
| Gene Symbol: CLGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130675.1 | NP_001124147.1 | 13 - 14 | 1701 | 151 | |
| NM_004362.2 | NP_004353.1 | 12 - 13 | 1617 | 151 | ||
| GenBank mRNA | AK093096.1 | - | 12 - 13 | 2180 | 151 | |
| AK290819.1 | - | 12 - 13 | 1614 | 151 | ||
| AK299243.1 | - | 11 - 12 | 1406 | 151 | ||
| AK302149.1 | - | 8 - 9 | 989 | 151 | ||
| BC028357.2 | - | 12 - 13 | 1571 | 4830135 | 151 | |
| D86322.1 | - | 12 - 13 | 1584 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001589.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | ENST00000325617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | ENST00000414773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | ENST00000537281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | ENST00000545667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022139.hg.1 | TC04001589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | ENST00000325617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | ENST00000414773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | ENST00000537281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | ENST00000545667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022138.hg.1 | TC04001589.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182098.hg.1 | TC0400011973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182099.hg.1 | TC0400011973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |

CLGN
Genomic Map