Assay Details
| Gene Symbol: CLGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130675.1 | NP_001124147.1 | 2 - 3 | 205 | 87 | |
| GenBank mRNA | AK093096.1 | - | 1 - 2 | 684 | 87 | |
| AL045602.1 | - | 2 - 3 | 149 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK093096.1, NM_001130675.1 transcripts.
Set Membership: |
|

CLGN
Genomic Map