Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000584.3 | NP_000575.1 | 3 - 4 | 444 | 142 | |
| GenBank mRNA | AK131067.1 | - | 3 - 4 | 381 | 142 | |
| BC013615.1 | - | 3 - 4 | 331 | 3882471 | 142 | |
| M17017.1 | - | 3 - 4 | 381 | 142 | ||
| M26383.1 | - | 3 - 4 | 365 | 142 | ||
| Y00787.1 | - | 3 - 4 | 392 | 142 | ||
More Information
Set Membership: |
|

CXCL8
Genomic Map