Assay Details
| Gene Symbol: SLC8A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058240.3 | NP_489479.1 | 4 - 5 | 2654 | 75 | |
| GenBank mRNA | AJ304853.1 | - | 3 - 4 | 1962 | 75 | |
More Information
Set Membership: |
|

SLC8A3
Genomic Map