Assay Details
| Gene Symbol: SLC8A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183002.2 | NP_892114.1 | 4 - 5 | 2655 | 81 | |
| XM_006720240.3 | XP_006720303.1 | 3 - 4 | 1501 | 81 | ||
| XM_017021606.1 | XP_016877095.1 | 4 - 5 | 2027 | 81 | ||
| XM_017021607.1 | XP_016877096.1 | 4 - 5 | 2520 | 81 | ||
| GenBank mRNA | AF510502.1 | - | 4 - 5 | 2655 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006720240.3 transcripts.
Set Membership: |
|

SLC8A3
Genomic Map