Assay Details
| Gene Symbol: ATG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286106.1 | NP_001273035.1 | 3 - 4 | 451 | 130 | |
| NM_001286108.1 | NP_001273037.1 | 3 - 4 | 586 | 130 | ||
| NM_004849.3 | NP_004840.1 | 3 - 4 | 586 | 130 | ||
| NR_104402.1 | 2 - 3 | 291 | 130 | |||
| GenBank mRNA | AK001899.1 | - | 3 - 4 | 553 | 130 | |
| AK315800.1 | - | 3 - 4 | 424 | 130 | ||
| BC002699.2 | - | 3 - 4 | 432 | 3609927 | 130 | |
| BC093011.1 | - | 3 - 4 | 513 | 30345897 | 130 | |
| EU283338.1 | - | 2 - 3 | 232 | 130 | ||
| EU283339.1 | - | 2 - 3 | 233 | 130 | ||
| HY168998.1 | - | 3 - 4 | 454 | 130 | ||
| JQ918353.1 | - | 2 - 3 | 233 | 130 | ||
| JQ924061.1 | - | 2 - 3 | 233 | 130 | ||
| Y11588.1 | - | 3 - 4 | 559 | 130 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001982.hg.1
Set Membership: |
|

ATG5
Genomic Map