Assay Details
| Gene Symbol: MYO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004999.3 | NP_004990.3 | 30 - 31 | 3459 | 71 | |
| XM_005248719.3 | XP_005248776.1 | 31 - 32 | 3502 | 71 | ||
| XM_005248720.3 | XP_005248777.1 | 31 - 32 | 3502 | 71 | ||
| XM_005248722.3 | XP_005248779.1 | 30 - 31 | 3475 | 71 | ||
| GenBank mRNA | AB002387.1 | - | 30 - 31 | 3319 | 71 | |
| AK095294.1 | - | 2 - 3 | 1373 | 71 | ||
| BC146764.1 | - | 30 - 31 | 3319 | 9007206 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

MYO6
Genomic Map