Assay Details
| Gene Symbol: TTC39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270644.2 | XP_005270701.1 | 1 - 2 | 154 | 72 | |
| XM_006710471.2 | XP_006710534.1 | 1 - 2 | 146 | 72 | ||
| GenBank mRNA | AB007921.1 | - | 1 - 2 | 171 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005270644.2, XM_006710471.2 transcripts.
Set Membership: |
|

TTC39A
Genomic Map