Assay Details
| Gene Symbol: FAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007051.2 | NP_008982.1 | 6 - 7 | 1007 | 79 | |
| XM_017000136.1 | XP_016855625.1 | 7 - 8 | 724 | 79 | ||
| GenBank mRNA | AF106798.1 | - | 6 - 7 | 554 | 79 | |
| AF132938.1 | - | 6 - 7 | 603 | 79 | ||
| AF136173.1 | - | 6 - 7 | 603 | 79 | ||
| AJ271408.1 | - | 6 - 7 | 879 | 79 | ||
| AK292840.1 | - | 7 - 8 | 617 | 79 | ||
| AL133631.1 | - | 3 - 4 | 267 | 79 | ||
| BC004970.1 | - | 6 - 7 | 1007 | 2958207 | 79 | |
| BC067100.1 | - | 6 - 7 | 831 | 5928559 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002644.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
cytoplasmic sequestering of NF-kappaB
cell death
positive regulation of cell death
regulation of cell adhesion
positive regulation of protein complex assembly
regulation of protein catabolic process
positive regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of protein kinase activity
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
protein binding
protein kinase regulator activity
protein kinase binding
protein domain specific binding
heat shock protein binding
ubiquitin protein ligase binding
ubiquitin binding
NF-kappaB binding
apoptotic process
cytoplasmic sequestering of NF-kappaB
cell death
positive regulation of cell death
regulation of cell adhesion
positive regulation of protein complex assembly
regulation of protein catabolic process
positive regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of protein kinase activity
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
protein binding
protein kinase regulator activity
protein kinase binding
protein domain specific binding
heat shock protein binding
ubiquitin protein ligase binding
ubiquitin binding
NF-kappaB binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | ENST00000607815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041350.hg.1 | ENST00000371778.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041350.hg.1 | ENST00000396153.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041350.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041350.hg.1 | TC01002644.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000371778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000472808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | ENST00000607815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014152.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | ENST00000396153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | ENST00000487898.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | ENST00000494400.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | NM_007051.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205833.hg.1 | TC0100014152.hg.1 | Clariom D TC |

FAF1
Genomic Map