Assay Details
| Gene Symbol: ALG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077691.1 | NP_001071159.1 | 10 - 11 | 1549 | 81 | |
| NM_024740.2 | NP_079016.2 | 10 - 11 | 1272 | 81 | ||
| XM_005277723.4 | XP_005277780.1 | 10 - 11 | 1272 | 81 | ||
| XM_006718913.2 | XP_006718976.1 | 10 - 11 | 1272 | 81 | ||
| XM_011542992.1 | XP_011541294.1 | 10 - 11 | 1272 | 81 | ||
| XM_017018314.1 | XP_016873803.1 | 9 - 10 | 1149 | 81 | ||
| XM_017018316.1 | XP_016873805.1 | 10 - 11 | 1569 | 81 | ||
| XM_017018319.1 | XP_016873808.1 | 10 - 11 | 1406 | 81 | ||
| XM_017018320.1 | XP_016873809.1 | 10 - 11 | 1276 | 81 | ||
| GenBank mRNA | AK025498.1 | - | 10 - 11 | 1272 | 81 | |
| AK172828.1 | - | 10 - 11 | 1133 | 81 | ||
| BC009255.2 | - | 10 - 11 | 1221 | 3029372 | 81 | |
| KU178783.1 | - | 10 - 11 | 1174 | 81 | ||
More Information
Set Membership: |
|

ALG9
Genomic Map