| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
ENST00000297405.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
ENST00000343508.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
ENST00000352409.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
ENST00000455883.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
NM_052900.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
NM_198123.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
NM_198124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020204.hg.1 |
TC08001538.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000297405.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000339701.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000343508.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000455883.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
NM_052900.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
NM_198123.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
NM_198124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000297405.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000339701.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000343508.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
ENST00000455883.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
NM_052900.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
NM_198123.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
NM_198124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178314.hg.1 |
TC0800011509.hg.1 |
Clariom D TC |