Assay Details
| Gene Symbol: AFG3L1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003227.1 | 6 - 7 | 695 | 89 | ||
| GenBank mRNA | AK097044.1 | - | 6 - 7 | 682 | 89 | |
| AK126911.1 | - | 5 - 6 | 659 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CENPB DNA-binding domain containing 1 | CENPBD1 | -11.7kb | Anti-sense | Chr. 16 - 89969775 - 89972832 on NCBI Build GRCh38 | |
| Downstream | dysbindin domain containing 1 | DBNDD1 | 13.9kb | Anti-sense | Chr. 16 - 90004865 - 90020131 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

AFG3L1P
Genomic Map