Assay Details
| Gene Symbol: CWF19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC028043.2 | - | 1 - 2 | 79 | 144 | |
| BC110439.1 | - | 12 - 13 | 1920 | 40001346 | 144 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC028043.2 transcripts.
Set Membership: |
|

CWF19L2
Genomic Map