Assay Details
| Gene Symbol: NR2F1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109818.1 | 1 - 2 | 184 | 68 | ||
| GenBank mRNA | AK095719.1 | - | 1 - 2 | 162 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2F1-AS1| Gene Symbol: NR2F1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109818.1 | 1 - 2 | 184 | 68 | ||
| GenBank mRNA | AK095719.1 | - | 1 - 2 | 162 | 68 | |
Set Membership: |
|