| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017161.hg.1 |
TC09001375.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017161.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017161.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017159.hg.1 |
TC09001375.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017159.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017159.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017160.hg.1 |
TC09001375.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017160.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017160.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017162.hg.1 |
TC09001375.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09017162.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174259.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174259.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174259.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174259.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174259.hg.1 |
TC0900010913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900174257.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174257.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174257.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174257.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174257.hg.1 |
TC0900010913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900174258.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174258.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174258.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174258.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174258.hg.1 |
TC0900010913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900174260.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174260.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174260.hg.1 |
ENST00000375256.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900174260.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174260.hg.1 |
TC0900010913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900174261.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174261.hg.1 |
NM_153695.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900174261.hg.1 |
TC0900010913.hg.1 |
Clariom D TC |