Assay Details
| Gene Symbol: MAD2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002358.3 | NP_002349.1 | 3 - 4 | 464 | 95 | |
| GenBank mRNA | AJ000186.1 | - | 3 - 4 | 406 | 95 | |
| AK223433.1 | - | 3 - 4 | 425 | 95 | ||
| AK313827.1 | - | 3 - 4 | 426 | 95 | ||
| BC000356.2 | - | 3 - 4 | 414 | 2964388 | 95 | |
| BC005945.1 | - | 3 - 4 | 407 | 4132892 | 95 | |
| BC070283.1 | - | 3 - 4 | 376 | 6654207 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MAD2L1
Genomic Map