| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014831.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014831.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014831.hg.1 |
TC11001173.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014833.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014833.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014833.hg.1 |
TC11001173.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014832.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014832.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014832.hg.1 |
TC11001173.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100171091.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171091.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171091.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171091.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171091.hg.1 |
TC1100009450.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171092.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171092.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171092.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171092.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171092.hg.1 |
TC1100009450.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100171089.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171089.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171089.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100171089.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100171089.hg.1 |
TC1100009450.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000263579.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000529149.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
ENST00000530860.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009450.hg.1 |
NM_014026.5 |
Refseq |