| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
TC1700006860.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700148362.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000262441.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000304773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000458005.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
ENST00000574745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006860.hg.1 |
NM_004246.2 |
Refseq |