Assay Details
| Gene Symbol: RIOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031480.2 | NP_113668.2 | 13 - 14 | 1441 | 130 | |
| NM_153005.1 | NP_694550.1 | 6 - 7 | 751 | 130 | ||
| XM_005249446.3 | XP_005249503.1 | 13 - 14 | 1486 | 130 | ||
| XM_011514933.2 | XP_011513235.2 | 13 - 14 | 1403 | 130 | ||
| GenBank mRNA | AF258661.1 | - | 6 - 7 | 751 | 130 | |
| AK054568.1 | - | 13 - 14 | 1432 | 130 | ||
| AK314467.1 | - | 13 - 14 | 1470 | 130 | ||
| AL834277.1 | - | 12 - 13 | 1140 | 130 | ||
| BC006104.1 | - | 13 - 14 | 1441 | 3537086 | 130 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000058.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000535.hg.1 | ENST00000379834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000535.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000535.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000535.hg.1 | TC06000058.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146984.hg.1 | TC0600006864.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |

RIOK1
Genomic Map