Assay Details
| Gene Symbol: RIOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031480.2 | NP_113668.2 | 3 - 4 | 542 | 129 | |
| XM_005249446.3 | XP_005249503.1 | 3 - 4 | 587 | 129 | ||
| XM_011514933.2 | XP_011513235.2 | 3 - 4 | 504 | 129 | ||
| GenBank mRNA | AK054568.1 | - | 3 - 4 | 533 | 129 | |
| AK314467.1 | - | 3 - 4 | 571 | 129 | ||
| AL834277.1 | - | 2 - 3 | 241 | 129 | ||
| AY094356.1 | - | 3 - 4 | 610 | 129 | ||
| BC006104.1 | - | 3 - 4 | 542 | 3537086 | 129 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000058.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005249446.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000518.hg.1 | ENST00000379834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000518.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000518.hg.1 | TC06000058.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000264874.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000484626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | ENST00000493691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006864.hg.1 | NM_153005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | ENST00000379834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | ENST00000475351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | NM_031480.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600146965.hg.1 | TC0600006864.hg.1 | Clariom D TC |

RIOK1
Genomic Map